missing translation for 'onlineSavingsMsg'
Learn More

AH 6809, MedChemExpress

Product Code. 16450317
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16450317

Brand: Medchem Express HY10418

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AH 6809 is an antagonist of EP and DP receptor, with Kis of 1217, 1150, 1597, and 1415 nM for the cloned human EP1, EP2, EP3-III, and DP receptor respectively. AH 6809 has a Ki of 350 nM for mouse EP2 receptor.

AH 6809 is an antagonist of EP and DP receptor, with Kis of 1217, 1150, 1597, and 1415 nM for the cloned human EP1, EP2, EP3-III, and DP receptor respectively. AH 6809 has a Ki of 350 nM for mouse EP2 receptor.

Specifications

Chemical Name or Material AH 6809
CAS 33458-93-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H14O5
Quantity 50 mg
Solubility Information DMSO : 25 mg/mL (83.81 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.34 mM; Need ultrasonic)
SMILES O=C1C2=C(C=CC(C(O)=O)=C2)OC3=CC(OC(C)C)=CC=C31
Molecular Weight (g/mol) 298.29
Formula Weight 298.29
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.