missing translation for 'onlineSavingsMsg'
Learn More

AGXT Antibody (2T4), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18429714 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429714 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429714 Supplier Novus Biologicals Supplier No. NBP144376MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

AGXT Monoclonal antibody specifically detects AGXT in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGXT
Applications Flow Cytometry
Classification Monoclonal
Clone 2T4
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias AGT1L-alanine: glyoxylate aminotransferase 1, AGThepatic peroxisomal alanine:glyoxylate aminotransferase, AGXT1, alanine-glyoxylate aminotransferase, Alanine--glyoxylate aminotransferase, EC 2.6.1.44, EC 2.6.1.51, PH1, serine:pyruvate aminotransferase, SPATserine-pyruvate aminotransferase, SPTserine--pyruvate aminotransferase, TLH6
Host Species Mouse
Immunogen Recombinant AGXT (330-392aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 189
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.