missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ AGXT Antibody (2D10), Novus Biologicals™

Mouse Monoclonal Antibody

£148.00 - £343.00

Specifications

Antigen AGXT
Clone 2D10
Concentration 1 mg/mL
Applications Flow Cytometry, ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30058543
View Documents
Novus Biologicals™
NBP3-31960-50ul
50 μL
£148.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30058517
View Documents
Novus Biologicals™
NBP3-31960-100ul
100 μL
£343.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AGXT Monoclonal antibody specifically detects AGXT in Human samples. It is validated for Flow Cytometry,ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

AGXT
1 mg/mL
Monoclonal
Purified
RUO
Human
AGT1L-alanine: glyoxylate aminotransferase 1, AGThepatic peroxisomal alanine:glyoxylate aminotransferase, AGXT1, alanine-glyoxylate aminotransferase, Alanine--glyoxylate aminotransferase, EC 2.6.1.44, EC 2.6.1.51, PH1, serine:pyruvate aminotransferase, SPATserine-pyruvate aminotransferase, SPTserine--pyruvate aminotransferase, TLH6
Recombinant AGXT (330-392aa) purified from E. coli
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
2D10
Flow Cytometry, ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Unconjugated
Mouse
Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction
PBS (pH 7.4), 10% Glycerol
189
IgG2b κ
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.