missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGTR-1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 8 publications
Brand: Novus Biologicals NBP1-77078
This item is not returnable.
View return policy
Description
AGTR-1 Polyclonal specifically detects AGTR-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| AGTR-1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| AG2S, AGTR1AAngiotensin II type-1 receptor, Angiostensin Receptor, angiotensin II receptor, type 1, angiotensin receptor 1, angiotensin receptor 1B, AT1AT1AR, AT1B, AT1R, AT2R1A, AT2R1AGTR1B, AT2R1BAT1BR, HAT1R, type-1 angiotensin II receptor, type-1B angiotensin II receptor | |
| Rabbit | |
| 41 kDa | |
| 0.1 mg | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry 2.5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin 2.5 ug/ml | |
| D3DNG8 | |
| AGTR1 | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the center of human AGTR1. The immunogen is located within amino acids 210 - 260 of AGTR1. Amino Acid Squence: KKAYEIQKNKPRNDDI | |
| Affinity Purified | |
| RUO | |
| 185 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction