missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AgRP Polyclonal Antibody
GREENER_CHOICE

Product Code. 10150724
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10150724 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10150724 Supplier Invitrogen™ Supplier No. PA118414

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Guinea Pig Polyclonal Antibody

Reconstitute in 50 μL sterile water. Centrifuge to remove any insoluble material. Store lyophilized antibody at 2-8 °C. It is recommended that a thawed sample is stored at 4 °C for no longer than 2 weeks. Allocation of appropriate anti-bacterial agent can increase shelf life by several weeks. Diluted serum should be prepared as required. Long term stability requires storage, preferably in small aliquots at -20 °C or lower. Glycerol (1:1) can be added to neat serum for additional stability if intended use does not prevent this.

Agouti-related protein (AgRP), also called agouti-related peptide, is a neuropeptide produced in the brain by the AgRP/NPY neuron. In humans, the agouti-related peptide is encoded by the AGRP gene. This gene encodes an antagonist of the melanocortin-3 and melanocortin-4 receptor. It appears to regulate hypothalamic control of feeding behavior via melanocortin receptor and/or intracellular calcium regulation, and thus plays a role in weight homeostasis. Mutations in this gene have been associated with late on-set obesity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AgRP
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with no preservative
Gene Agrp
Gene Accession No. O00253, P56473
Gene Alias agouti related neuropeptide; agouti related protein; agouti related protein homolog; agouti-related protein; Agrp; AGRT; ART; ASIP2; orexigenic neuropeptide
Gene Symbols Agrp
Host Species Guinea Pig
Immunogen Synthetic peptide corresponding to a region (82-131) within the carboxy domain of mouse agouti related protein conjugated to carrier protein.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11604, 181, 25582, 443269
Target Species Human, Mouse, Rat, Sheep
Content And Storage 4°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.