Learn More
Invitrogen™ AgRP Human ELISA Kit
Sandwich ELISA Kit
£546.00 - £2310.00
Specifications
Accession Number | O00253 |
---|---|
Assay Sensitivity | 6 pg/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15424893
|
Invitrogen™
EHAGRP |
96 Tests |
£546.00
1 set |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17175339
|
Invitrogen™
EHAGRPX5 |
5 x 96 Tests |
£2310.00
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
Description
The Human Agouti Related Peptide (Hu AgRP) ELISA quantitates Hu AgRP in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu AgRP. Principle of the method The Human AgRP solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
AgRP plays a role in weight homeostasis. Involved in the control of feeding behavior through the central melanocortin system. Acts as alpha melanocyte-stimulating hormone antagonist by inhibiting cAMP production mediated by stimulation of melanocortin receptors within the hypothalamus and adrenal gland. Has very low activity with MC5R (By similarity). Is an inverse agonist for MC3R and MC4R being able to suppress their constitutive activity. It promotes MC3R and MC4R endocytosis in an arrestin-dependent manner.Specifications
O00253 | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
Human | |
Colorimetric Microplate Reader | |
AGRT,ART,ASIP2 | |
<10% | |
RUO | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
6 pg/mL | |
Biotin | |
Other Proteins | |
This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, CNTF, ENA- 78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL- 11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, FGF-4, FGF-6, FGF- 7, G-CSF, GDNF, GM-CSF, IFN-gamma, IGFBP-2, IGFBP-3, IGFBP-4, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIF, MIG, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, SDF-1 alpha, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF. | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
181 | |
<12% | |
HRP | |
Neurobiology | |
AGRT, ART, ASIP2 | |
1 hr. 20 min. |