missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AgRP/ART Antibody (MM0085-32B12), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12026PE
This item is not returnable.
View return policy
Description
AgRP/ART Monoclonal antibody specifically detects AgRP/ART in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| AgRP/ART | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0085-32B12 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| agouti related protein homolog (mouse), agouti-related protein, Agrt, AGRTMGC118963, ARTagouti (mouse) related protein, ASIP2 | |
| Human recombinant AgRP/ART | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 181 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction