missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Agrin Monoclonal Antibody (76805)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561564

Product Code. 30534163

  • £490.00 / 500µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

Agrin is a molecule that resides in the basal lamina of muscle cells and directs key events in post synaptic differentiation. Most notably, Agrin is responsible for the clustering of acetylcholine receptors (AChRs) on the cell surface and their localization to the neuromuscular junction. Several Agrin variants have been identified which arise from alternative mRNA splicings. Agrin splice forms having inserts at two sites in the carboxy terminus designated 'y' and 'z' display a high affinity for AChRs, while splice forms lacking these inserts associate with AChRs weakly. Muscle alpha-dystroglycan has been postulated to be the receptor for the clustering activity of agrin; however, this is a point of contention. Tyrosine phosphorylation has been implicated as a required early step in AChR aggregation. Interestingly, a unique receptor tyrosine kinase, designated MuSK, has been discovered that interacts with Agrin and is specifically localized to developing muscle.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Agrin
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
P25304
Agrn
S. frugiperda insect ovarian cell line Sf 21-derived recombinant rat Agrin, Ala1153-Pro1959 (Pro1788-Ser1798 del).
500 μg
Primary
Rat
Antibody
IgG1
Western Blot
76805
Unconjugated
Agrn
AGR; Agrin; Agrin C-terminal 110 kDa subunit; Agrin C-terminal 22 kDa fragment; Agrin C-terminal 90 kDa fragment; Agrin N-terminal 110 kDa subunit; agrin proteoglycan; Agrn; C22; C90; CMS8; CMSPPD; nmf380; RP23-139J18.2
Mouse
Protein A/G
RUO
25592
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.