missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGR2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12275-1-AP-150UL
This item is not returnable.
View return policy
Description
AGR2 (anterior gradient protein 2) and AGR3 are members of the anterior gradient homolog family. They are secreted cytoplasmic proteins which are involved in metastasis induction and p53 tumour supressor inhibition. They may serve as molecular markers and potential therapeutic targets for hormone-responsive breast tumours. AGR2 is ubiquitously expressed with upregulated expression in prostate cancer, breast cancer, lung cancer, renal carcinomas and endometrial carcinomas. AGR2 expression is positively correlated with that of the estrogen receptor (ER) and is negatively correlated with that of the epidermal growth factor receptor (EGFR).Specifications
| AGR2 | |
| Polyclonal | |
| Unconjugated | |
| AGR2 | |
| AG 2, AG2, AGR2, GOB 4, HAG 2, HPC8, PDIA17, XAG 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10551, 23795, 298961 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O88312, O95994 | |
| AGR2 | |
| AGR2 Fusion Protein Ag2919 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction