missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGR2 Mouse anti-Human, Porcine, Clone: 1A8A8, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | AGR2 |
|---|---|
| Clone | 1A8A8 |
| Concentration | 0.93 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16888945
|
Proteintech
66768-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16878945
|
Proteintech
66768-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AGR2 (anterior gradient protein 2) and AGR3 are members of the anterior gradient homolog family. They are secreted cytoplasmic proteins which are involved in metastasis induction and p53 tumour supressor inhibition. They may serve as molecular markers and potential therapeutic targets for hormone-responsive breast tumours. AGR2 is ubiquitously expressed with upregulated expression in prostate cancer, breast cancer, lung cancer, renal carcinomas and endometrial carcinomas. AGR2 expression is positively correlated with that of the estrogen receptor (ER) and is negatively correlated with that of the epidermal growth factor receptor (EGFR).Specifications
| AGR2 | |
| 0.93 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| AG 2, AG2, AGR2, GOB 4, HAG 2, HPC8, PDIA17, XAG 2 | |
| AGR2 | |
| IgG2b | |
| Protein A | |
| Antibody |
| 1A8A8 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Pig | |
| O95994 | |
| 100514482, 10551 | |
| AGR2 Fusion Protein Ag2919 | |
| Primary | |
| -20°C | |
| AGR2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title