missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AGPS Monoclonal Antibody (AGPS-03)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA528603
This item is not returnable.
View return policy
Description
This antibody recognizes AGPS (alkykglycerone phosphate synthase), an intracellular peroxisomal enzyme important for lipid biosynthesis.
This gene is a member of the FAD-binding oxidoreductase/transferase type 4 family. It encodes a protein that catalyzes the second step of ether lipid biosynthesis in which acyl-dihydroxyacetonephosphate (DHAP) is converted to alkyl-DHAP by the addition of a long chain alcohol and the removal of a long-chain acid anion. The protein is localized to the inner aspect of the peroxisomal membrane and requires FAD as a cofactor. Mutations in this gene have been associated with rhizomelic chondrodysplasia punctata, type 3 and Zellweger syndrome.
Specifications
| AGPS | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| O00116 | |
| AGPS | |
| recombinant human AGPS (amino acids 158-384). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Western Blot | |
| AGPS-03 | |
| Unconjugated | |
| AGPS | |
| 5832437L22; 9930035G10Rik; AAG5; Adaps; Adap-s; ADAS; Adhaps; ADPS; Ads; Aging-associated gene 5 protein; aging-associated protein 5; AGPS; ALDHPSY; alkyl-DHAP synthase; alkyldihydroxyacetone phosphate synthase; alkyl-dihydroxyacetonephosphate synthase; alkyldihydroxyacetonephosphate synthase, peroxisomal; alkylglycerone phosphate synthase; alkylglycerone phosphate synthase-like; alkylglycerone-phosphate synthase; AW123847; bs2; RCDP3 | |
| Mouse | |
| Protein A | |
| RUO | |
| 8540 | |
| 4°C, do not freeze | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction