missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AGPAT6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553428
This item is not returnable.
View return policy
Description
Immunogen sequence: FAWATLRMER GAKEKNHQLY KPYTNGIIAK DPTSLEEEIK EIRRSGSSKA LDNTPEFELS DIFYFCRKGM ETIMDDEVTK RFSAEELESW Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
Lysophosphatidic acid acyltransferases (EC 2.3.1.51) catalyze the conversion of lysophosphatidic acid (LPA) to phosphatidic acid (PA). LPA and PA are involved in signal transduction and lipid biosynthesis.
Specifications
| AGPAT6 | |
| Polyclonal | |
| Unconjugated | |
| GPAT4 | |
| 1-acylglycerol-3-phosphate O-acyltransferase 6; 1-acylglycerol-3-phosphate O-acyltransferase 6 (lysophosphatidic acid acyltransferase, zeta); 1-acyl-sn-glycerol-3-phosphate acyltransferase zeta; 1-AGP; 1-AGP acyltransferase 6; 1-AGPAT 6; acyl-CoA:glycerol-3-phosphate acyltransferase 4; AGPAT6; AU041707; AW545732; Glycerol-3-phosphate acyltransferase 4; glycerol-3-phosphate acyltransferase 6; glycerol-3-phosphate acyltransferase-4; Gpat4; LPAATZ; LPAAT-zeta; lysophosphatidic acid acyltransferase zeta; putative lysophosphatidic acid acyltransferase; RGD1310520; testis spermatogenesis apoptosis-related protein 7; TSARG7; UNQ551/PRO1108 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 137964 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86UL3 | |
| GPAT4 | |
| Recombinant protein corresponding to Human AGPAT6. Recombinant protein control fragment (Product #RP-91086). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction