missing translation for 'onlineSavingsMsg'
Learn More

AGPAT6 Antibody, Novus Biologicals™

Product Code. 18499850 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18499850 25 μL 25µL
18428750 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18499850 Supplier Novus Biologicals Supplier No. NBP18506125ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AGPAT6 Polyclonal antibody specifically detects AGPAT6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGPAT6
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias 1-acylglycerol-3-phosphate O-acyltransferase 6 (lysophosphatidic acidacyltransferase, zeta), Acyl-CoA:glycerol-3-phosphate acyltransferase 4, DKFZp586M1819,1-acyl-sn-glycerol-3-phosphate acyltransferase zeta, EC 2.3.1.15, glycerol-3-phosphate acyltransferase 4,1-AGP acyltransferase 6, glycerol-3-phosphate acyltransferase 6,1-AGPAT 6, GPAT4,1-acylglycerol-3-phosphate O-acyltransferase 6, LPAAT-zetaLPAATZ, Lysophosphatidic acid acyltransferase zeta, testis spermatogenesis apoptosis-related protein 7, TSARG7
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: FAWATLRMERGAKEKNHQLYKPYTNGIIAKDPTSLEEEIKEIRRSGSSKALDNTPEFELSDIFYFCRKGMETIMDDEVTKRFSAEELESW
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Diabetes Research, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 137964
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.