missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGPAT6 Antibody (4C12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00137964-M02
This item is not returnable.
View return policy
Description
AGPAT6 Monoclonal antibody specifically detects AGPAT6 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| AGPAT6 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| 1-acylglycerol-3-phosphate O-acyltransferase 6 (lysophosphatidic acidacyltransferase, zeta), Acyl-CoA:glycerol-3-phosphate acyltransferase 4, DKFZp586M1819,1-acyl-sn-glycerol-3-phosphate acyltransferase zeta, EC 2.3.1.15, glycerol-3-phosphate acyltransferase 4,1-AGP acyltransferase 6, glycerol-3-phosphate acyltransferase 6,1-AGPAT 6, GPAT4,1-acylglycerol-3-phosphate O-acyltransferase 6, LPAAT-zetaLPAATZ, Lysophosphatidic acid acyltransferase zeta, testis spermatogenesis apoptosis-related protein 7, TSARG7 | |
| AGPAT6 (NP_848934.1, 39 a.a. ∽ 137 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SFGIRKLYMKSLLKIFAWATLRMERGAKEKNHQLYKPYTNGIIAKDPTSLEEEIKEIRRSGSSKALDNTPEFELSDIFYFCRKGMETIMDDEVTKRFSA | |
| 0.1 mg | |
| Lipid and Metabolism | |
| 137964 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| 4C12 | |
| Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence | |
| NP_848934 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction