missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGPAT3 Polyclonal Antibody

AGPAT3 Polyclonal Antibody
Brand: Invitrogen PA549623
This item is not returnable.
View return policy
Description
AGPAT3 Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
| AGPAT3 | |
| Polyclonal | |
| Unconjugated | |
| Agpat3 | |
| 1-acylglycerol-3-phosphate O-acyltransferase 3; 1-acyl-sn-glycerol-3-phosphate acyltransferase gamma; 1-AGP acyltransferase 3; 1-AGPAT 3; Agpat3; AW061257; AW493985; D10Jhu12e; lpaat3; LPAAT-gamma; LPAAT-GAMMA1; lysophosphatidic acid acyltransferase 3; lysophosphatidic acid acyltransferase gamma; lysophosphatidic acid acyltransferase-gamma1; UNQ759/PRO1490 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 56894 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9NRZ7 | |
| Agpat3 | |
| KLH conjugated synthetic peptide between 241-269 amino acids from the Central region of human AGPAT3. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction