missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AGPAT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112352
This item is not returnable.
View return policy
Description
AGPAT2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Lysophospholipid acyltransferases (LPLATs) catalyse the addition of fatty acyl moieties to the glycerol backbone of lysophospholipids. In addition to playing a crutial role in the synthesis of structural membrane components the LPLATs are also implicated in cellular signalling responses for cytokines, growth factors and other agonists. Cellular signalling through the interleukin 1 receptor in human mesangial cells and EL-4 cells proceeds by the activation of lysophosphatidic acid acyltransferase (LPAAT). Activation of LPAAT by interleukin 1 results in the generation of unsaturated phosphatidic acid species, that are crucial to the generation of diacylglycerol and interleukin 1 signalling. LPLATs are also involved in signalling for increased interleukin 2 synthesis through the T cell antigen receptor. Activation of T-cells via anti-CD3 stimulation has been shown to increase incorporation of polyunsaturated fatty acids intophosphatidylcholine via lysophosphatidylcholine acyltransferase.Activation of this enzyme is essential for the sustained activationand translocation of protein kinase C.
Specifications
| AGPAT2 | |
| Polyclonal | |
| Unconjugated | |
| AGPAT2 | |
| 1-acylglycerol-3-phosphate O-acyltransferase 2; 1-acylglycerol-3-phosphate O-acyltransferase 2 (lysophosphatidic acid acyltransferase, beta); 1-acyl-sn-glycerol-3-phosphate acyltransferase beta; 1-AGP acyltransferase 2; 1-AGPAT 2; 1-AGPAT2; 2510002J07Rik; AGPAT2; AV000834; BSCL; BSCL1; LPAAB; LPAAT-beta; lysophosphatidic acid acyltransferase beta; lysophosphatidic acid acyltransferase-beta; testicular tissue protein Li 143 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10555 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.55 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O15120 | |
| AGPAT2 | |
| Recombinant Human 1-acyl-sn-glycerol-3-phosphate acyltransferase beta protein (199-278AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction