missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGPAT1, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00010554-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MDLWPGAWMLLLLLFLLLLFLLPTLWFCSPSAKYFFKMAFYNGWILFLAVLAIPVCAVRGRNVENMKILRLMLLHIKYLYGIRVEVRGAHHFPPSQPYVVVSNHQSSLDLLGMMEVLPGRCVPIAKRELLWAGSAGLACWLAGVIFIDRKRTGDAISVMSEVAQTLLTQDVRVWVFPEGTRNHNGSMLPFKRGAFHLAVQAQVPIVPIVMSSYQDFYCKKERRFTSGQCQVRVLPPVPTEGLTPDDVPALADRVRHSMLTVFREISTDGRGGGDYLKKPGGGGSpecifications
| AGPAT1 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| BC002402 | |
| AGPAT1 | |
| AGPAT1 (AAH02402, 1 a.a. ∼ 283 a.a) full-length human protein. | |
| RUO | |
| 10554 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| 1-acylglycerol-3-phosphate O-acyltransferase 1 (lysophosphatidic acid acyltransferase, alpha) | |
| 1-AGPAT1/G15/LPAAT-alpha/LPAATA/MGC4007/MGC5423 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction