missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agomelatine (L(+)-Tartaric acid), MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
Agomelatine L(+)-Tartaric acid (S-20098 L(+)-Tartaric acid) is a specific agonist of MT1 and MT2 receptors with Kis of 0.1, 0.06, 0.12, and 0.27 nM for CHO-hMT1, HEK-hMT1, CHO-hMT2, and HEK-hMT2, respectively. Agomelatine L(+)-Tartaric acid is a selective 5-HT2C receptor antagonist with pKis of 6.4 and 6.2 at native (porcine) and cloned, human 5-HT2C receptors, respectively.
Specifications
Specifications
| Chemical Name or Material | Agomelatine (L(+)-Tartaric acid) |
| CAS | 824393-18-2 |
| Purity Grade Notes | Research |
| Molecular Formula | C19H23NO8 |
| Quantity | 10 mg |
| Synonym | S-20098 L(+)-Tartaric acid |
| Solubility Information | DMSO : 100 mg/mL (254.20 mM; Need ultrasonic) |
| SMILES | CC(NCCC1=C2C=C(OC)C=CC2=CC=C1)=O.O=C(O)[C@H](O)[C@@H](O)C(O)=O |
| Molecular Weight (g/mol) | 393.39 |
| Formula Weight | 393.39 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction