missing translation for 'onlineSavingsMsg'
Learn More

Agomelatine (L(+)-Tartaric acid), MedChemExpress

Product Code. 16479797
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16479797

Brand: Medchem Express HY17038B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Agomelatine L(+)-Tartaric acid (S-20098 L(+)-Tartaric acid) is a specific agonist of MT1 and MT2 receptors with Kis of 0.1, 0.06, 0.12, and 0.27 nM for CHO-hMT1, HEK-hMT1, CHO-hMT2, and HEK-hMT2, respectively. Agomelatine L(+)-Tartaric acid is a selective 5-HT2C receptor antagonist with pKis of 6.4 and 6.2 at native (porcine) and cloned, human 5-HT2C receptors, respectively.

Agomelatine L(+)-Tartaric acid (S-20098 L(+)-Tartaric acid) is a specific agonist of MT1 and MT2 receptors with Kis of 0.1, 0.06, 0.12, and 0.27 nM for CHO-hMT1, HEK-hMT1, CHO-hMT2, and HEK-hMT2, respectively. Agomelatine L(+)-Tartaric acid is a selective 5-HT2C receptor antagonist with pKis of 6.4 and 6.2 at native (porcine) and cloned, human 5-HT2C receptors, respectively.

Specifications

Chemical Name or Material Agomelatine (L(+)-Tartaric acid)
CAS 824393-18-2
Purity Grade Notes Research
Molecular Formula C19H23NO8
Quantity 5 mg
Synonym S-20098 L(+)-Tartaric acid
Solubility Information DMSO : 100 mg/mL (254.20 mM; Need ultrasonic)
SMILES CC(NCCC1=C2C=C(OC)C=CC2=CC=C1)=O.O=C(O)[C@H](O)[C@@H](O)C(O)=O
Molecular Weight (g/mol) 393.39
Formula Weight 393.39
Percent Purity 99.82%
Grade Research
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.