missing translation for 'onlineSavingsMsg'
Learn More

Agomelatine hydrochloride, MedChemExpress

Product Code. p-200101474
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493297 50 mg 50mg
16469797 5 mg 5mg
16427288 10 mg 10mg
16417246 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16493297 Supplier Medchem Express Supplier No. HY17038A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Agomelatine hydrochloride (S-20098 hydrochloride) is a specific agonist of MT1 and MT2 receptors with Kis of 0.1, 0.06, 0.12, and 0.27 nM for CHO-hMT1, HEK-hMT1, CHO-hMT2, and HEK-hMT2, respectively. Agomelatine hydrochloride is a selective 5-HT2C receptor antagonist with pKis of 6.4 and 6.2 at native (porcine) and cloned, human 5-HT2C receptors, respectively.

Agomelatine hydrochloride (S-20098 hydrochloride) is a specific agonist of MT1 and MT2 receptors with Kis of 0.1, 0.06, 0.12, and 0.27 nM for CHO-hMT1, HEK-hMT1, CHO-hMT2, and HEK-hMT2, respectively. Agomelatine hydrochloride is a selective 5-HT2C receptor antagonist with pKis of 6.4 and 6.2 at native (porcine) and cloned, human 5-HT2C receptors, respectively.

Specifications

Chemical Name or Material Agomelatine hydrochloride
CAS 1176316-99-6
Color White
Purity Grade Notes Research
Molecular Formula C15H18ClNO2
Quantity 50 mg
Synonym S-20098 hydrochloride
Solubility Information DMSO : ≥ 100 mg/mL (357.45 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC(NCCC1=C2C=C(OC)C=CC2=CC=C1)=O.Cl
Molecular Weight (g/mol) 279.76
Formula Weight 279.76
Percent Purity 99.55%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.