missing translation for 'onlineSavingsMsg'
Learn More

Agomelatine-d4, TRC

Product Code. 30322281
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30322281 1 mg 1mg
30321201 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30322281 Supplier Toronto Research Chemicals Supplier No. TRCA4300021MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Agomelatine-d4
CAS 1079389-44-8
Molecular Formula C15 D4 H13 N O2
Quantity 1 mg
Synonym N-[2-(7-Methoxy-1-naphthalenyl)ethyl-d4]acetamide, N-[2-(7-Methoxy-1-naphthyl) ethyl]acetamide-d4, S 20098-d4, Valdoxan-d4
InChI Formula InChI=1S/C15H17NO2/c1-11(17)16-9-8-13-5-3-4-12-6-7-14(18-2)10-15(12)13/h3-7,10H,8-9H2,1-2H3,(H,16,17)/i8D2,9D2
SMILES [2H]C([2H])(NC(=O)C)C([2H])([2H])c1cccc2ccc(OC)cc12
IUPAC Name N-[1,1,2,2-tetradeuterio-2-(7-methoxynaphthalen-1-yl)ethyl]acetamide
Molecular Weight (g/mol) 247.326
Formula Weight 247.151
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.