missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ago2/eIF2C2 Antibody (7C2), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | Ago2/eIF2C2 |
|---|---|
| Clone | 7C2 |
| Dilution | Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Flow Cytometry, ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043391
|
Novus Biologicals
NBP3-26520-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043643
|
Novus Biologicals
NBP3-26520-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ago2/eIF2C2 Monoclonal antibody specifically detects Ago2/eIF2C2 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunocytochemistrySpecifications
| Ago2/eIF2C2 | |
| Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AGO2argonaute 2, argonaute2, EC 3.1.26.n1, EC 3.1.26.n2, eIF2C 2, eIF-2C 2, Eukaryotic translation initiation factor 2C 2, eukaryotic translation initiation factor 2C, 2, hAgo2, MGC3183, PAZ Piwi domain protein, PPD, protein argonaute-2, Protein slicer, Q10 | |
| A synthesized peptide derived from Human Ago2/eIF2C2 [UniProt Q9UKV8] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 7C2 | |
| Flow Cytometry, ELISA, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Epigenetics | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 27161 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title