missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGO1/EIF2C1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92465-0.02ml
This item is not returnable.
View return policy
Description
AGO1/EIF2C1 Polyclonal antibody specifically detects AGO1/EIF2C1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| AGO1/EIF2C1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| AGO1Q99, argonaute 1, argonaute1, DKFZp686M13167, EIF2C, eIF2C 1, eIF-2C 1, Eukaryotic translation initiation factor 2C 1, eukaryotic translation initiation factor 2C, 1, GERP95, Golgi Endoplasmic Reticulum protein 95 kDa, hAgo1, protein argonaute-1, Putative RNA-binding protein Q99 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-170 of human AGO1/EIF2C1 (NP_036331.1). MEAGPSGAAAGAYLPPLQQVFQAPRRPGIGTVGKPIKLLANYFEVDIPKIDVYHYEVDIKPDKCPRRVNREVVEYMVQHFKPQIFGDRKPVYDGKKNIYTVTALPIGNERVDFEVTIPGEGKDRIFKVSIKWLAIVSWRMLHEALVSGQIPVPLESVQALDVAMRHLASM | |
| 0.02 mL | |
| Cell Biology, Cell Cycle and Replication, Immunology | |
| 26523 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction