missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AGO1/AGO2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540075
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
AGO1/AGO2 Polyclonal Antibody for Western Blot, ELISA
IEF2C1 and 2 are members of the Argonaute family. These proteins are involved in short-interfering-RNA-mediated gene silencing. This process is mediated by the RISC (RNA induced silencing complex), a multicomponent nuclease that is able to degrade double stranded RNAs. The mRNA degradation is triggered by the association of specific siRNAs with their homologous mRNAs. IEF2C2 has been shown to be essential for embryonic development.
Specifica
| AGO1/AGO2 | |
| Polyclonal | |
| Unconjugated | |
| AGO2 | |
| AGO1; AGO2; argonaute 1; argonaute 1, RISC catalytic component; argonaute 2; argonaute 2, RISC catalytic component; argonaute RISC catalytic component 1; Argonaute RISC catalytic component 2; argonaute1; Argonaute2; CTA-204B4.6; EIF2C; eIF2C 1; eIF-2C 1; eIF2C 2; eIF-2C 2; EIF2C1; EIF2C2; Eukaryotic translation initiation factor 2C 1; Eukaryotic translation initiation factor 2C 2; eukaryotic translation initiation factor 2C, 1; eukaryotic translation initiation factor 2C, 2; GERP95; Golgi Endoplasmic Reticulum protein 95 kDa; hAgo1; hAgo2; PAZ Piwi domain protein; PPD; protein argonaute-1; Protein argonaute-2; protein slicer; Putative RNA-binding protein Q99; Q10; Q99 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| Conc. Not Determined | |
| Whole serum with 0.05% sodium azide | |
| Q9UKV8, Q9UL18 | |
| AGO1, AGO2 | |
| KLH-conjugated synthetic peptide containing a sequence from the C-terminal part of both EIF2C1 and EIF2C2 corresponding to human IEF2C1/2 (eukaryotic translation initiation factor 2C1 and 2). | |
| RUO | |
| 26523, 27161 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto