missing translation for 'onlineSavingsMsg'
Learn More

AGO1/2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18372076 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18372076 25 μg 25µL
18302293 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18372076 Supplier Novus Biologicals Supplier No. NBP31865425UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AGO1/2 Polyclonal antibody specifically detects AGO1/2 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGO1/2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:100
Formulation Whole antiserum
Gene Accession No. Q9UL18
Gene Alias argonaute 1, RISC catalytic component, argonaute 2, RISC catalytic component, argonaute RISC catalytic component 1, argonaute1, cancer susceptibility candidate 7 (non-protein coding), CASC7, EIF2C, eIF2C 1, eIF-2C 1, EIF2C1, EIF2C2, eukaryotic translation initiation factor 2C, 1, eukaryotic translation initiation factor 2C, 2, GERP95, Golgi Endoplasmic Reticulum protein 95 kDa, hAgo1, LESKRES, LINC00980, long intergenic non-protein coding RNA 980, PAZ Piwi domain protein, PPD, protein argonaute-1, protein argonaute-2, Protein slicer, Putative RNA-binding protein Q99, Q10, Q99
Host Species Rabbit
Immunogen Polyclonal antibody raised in rabbit against human AGO1/2 (eukaryotic translation initiation factor 2C1 and 2), using a KLH-conjugated synthetic peptide containing a sequence from the C-terminal part of both EIF2C1 and EIF2C2.
Purification Method Whole Antiserum
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26523
Target Species Human
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Antisera
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.