missing translation for 'onlineSavingsMsg'
Learn More

AGN-195183, MedChemExpress

Product Code. 16439667
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16439667

Brand: Medchem Express HY16684

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AGN-195183 (IRX-5183) is a potent and selective agonist of RARα (Kd=3 nM) with improved binding selectivity relative to AGN 193836. AGN-195183 has no activity on RARβ/γ.

AGN-195183 (IRX-5183) is a potent and selective agonist of RARα (Kd=3 nM) with improved binding selectivity relative to AGN 193836. AGN-195183 has no activity on RARβ/γ.

Specifications

Chemical Name or Material AGN-195183
CAS 367273-07-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H22ClF2NO4
Quantity 5 mg
Synonym IRX-5183 VTP-195183 NRX-195183
Solubility Information DMSO : 11 mg/mL (25.12 mM; Need ultrasonic)
SMILES O=C(O)C1=C(F)C=C(NC(C2=C(O)C(Cl)=C3C(C)(C)CCC(C)(C)C3=C2)=O)C=C1F
Molecular Weight (g/mol) 437.86
Formula Weight 437.86
Percent Purity 98.1%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.