missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AGN 192403 hydrochloride
GREENER_CHOICE

Product Code. 18025762
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18025762 50 mg 50mg
18025472 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18025762 Supplier Tocris Bioscience™ Supplier No. 1072/50
  • ONLINE EXCLUSIVE PRICE
    £462.00
    £436.80 / 50mg
    Save £25.20
    5% Off
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

I1 selective ligand

I1 imidazoline binding site selective ligand with a potency at I1 comparable to moxonidine, but devoid of affinity for adrenoceptors and the I2 binding site. Interestingly, in animal models, this compound causes none of the physiological responses associated with the I1 binding site.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1021868-90-5
Molecular Formula C10H20ClN

Specifications

Quantity 50 mg
Formula Weight 189.73
Purity >98%
Molecular Weight (g/mol) 189.727
Chemical Name or Material AGN 192403 hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.