missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AGL Recombinant Rabbit Monoclonal Antibody (23GB2965), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553593
This item is not returnable.
View return policy
Description
This gene encodes the glycogen debrancher enzyme which is involved in glycogen degradation. This enzyme has two independent catalytic activities which occur at different sites on the protein: a 4-alpha-glucotransferase activity and a amylo-1,6-glucosidase activity. Mutations in this gene are associated with glycogen storage disease although a wide range of enzymatic and clinical variability occurs which may be due to tissue-specific alternative splicing. Alternatively spliced transcripts encoding different isoforms have been described.
Specifications
| AGL | |
| Recombinant Monoclonal | |
| Unconjugated | |
| AGL | |
| 1-Apr; 4-alpha-glucanotransferase; 4-glucantransferase; AGL; amylo-1, 6-glucosidase, 4-alpha-glucanotransferase; Amylo-1,6-glucosidase; amylo-alpha-1, 6-glucosidase, 4-alpha-glucanotransferase; amylo-alpha-1, 6-glucosidase, 4-alpha-glucanotransferaseprovided; Amylo-alpha-1,6-glucosidase; Dextrin 6-alpha-D-glucosidase; GDE; glycogen debrancher; Glycogen debranching enzyme; glycogen debranching protein; Oligo-1; Oligo-1,4-1,4-glucantransferase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 178, 362029, 77559 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2965 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P35573 | |
| AGL | |
| A synthesized peptide derived from AGL (60-90AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction