missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGL Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-91993-0.02ml
This item is not returnable.
View return policy
Description
AGL Polyclonal antibody specifically detects AGL in Human, Mouse samples. It is validated for Western Blot
Specifications
| AGL | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| amylo-alpha-1, 6-glucosidase, 4-alpha-glucanotransferase, GDEamylo-1, 6-glucosidase, 4-alpha-glucanotransferase, Glycogen debrancher, glycogen debranching enzyme | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1233-1532 of human AGL (NP_000633.2). EGFNITAGVDEETGFVYGGNRFNCGTWMDKMGESDRARNRGIPATPRDGSAVEIVGLSKSAVRWLLELSKKNIFPYHEVTVKRHGKAIKVSYDEWNRKIQDNFEKLFHVSEDPSDLNEKHPNLVHKRGIYKDSYGASSPWCDYQLRPNFTIAMVVAPELFTTEKAWKALEIAEKKLLGPLGMKTLDPDDMVYCGIYDNALDNDNYNLAKGFNYHQGPEWLWPIGYFLRAKLYFSRLMGPETTAKTIVLVKNVLSRHYVHLERSPWKGLPELTNENAQYCPFSCETQAWSIATILETLYDL | |
| 0.02 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 178 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction