missing translation for 'onlineSavingsMsg'
Learn More

AGL-2263, MedChemExpress

Product Code. 16417407
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16417407

Brand: Medchem Express HY112720

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AGL-2263 is an insulin receptor and insulin-like growth factor (IGF) receptor inhibitor.

AGL-2263 is an insulin receptor and insulin-like growth factor (IGF) receptor inhibitor.

Specifications

Chemical Name or Material AGL-2263
CAS 638213-98-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H10N2O5
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (387.87 mM; Need ultrasonic)
SMILES O=C(C1=CC(O)=C(O)C=C1)/C(C#N)=C/C2=CC=C(OC3=O)C(N3)=C2
Molecular Weight (g/mol) 322.27
Formula Weight 322.27
Percent Purity 97.04%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.