missing translation for 'onlineSavingsMsg'
Learn More

AGK2, MedChemExpress

Product Code. 16473088
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16473088 10 mg 10mg
16442616 25 mg 25mg
16425827 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16473088 Supplier Medchem Express Supplier No. HY100578
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress AGK2 is a selective SIRT2 inhibitor with an IC50 of 3.5 μM. AGK2 inhibits SIRT1 and SIRT3 with IC50s of 30 and 91 μM, respectively.

AGK2 is a selective SIRT2 inhibitor with an IC50 of 3.5 μM. AGK2 inhibits SIRT1 and SIRT3 with IC50s of 30 and 91 μM, respectively.

Specifications

Chemical Name or Material AGK2
CAS 304896-28-4
Color Green Yellow
Purity Grade Notes Research
Molecular Formula C23H13Cl2N3O2
Quantity 10 mg
Solubility Information DMSO : 12.5 mg/mL (28.78 mM; Need ultrasonic)
SMILES O=C(NC1=C2C=CC=NC2=CC=C1)/C(C#N)=C/C3=CC=C(C4=CC(Cl)=CC=C4Cl)O3
Molecular Weight (g/mol) 434.27
Formula Weight 434.27
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.