missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AGK Recombinant Rabbit Monoclonal Antibody (HL1846)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550341
This item is not returnable.
View return policy
Description
AGK Recombinant Monoclonal Antibody for Western Blot
Lipid kinase that can phosphorylate both monoacylglycerol and diacylglycerol to form lysophosphatidic acid (LPA) and phosphatidic acid (PA), respectively. Does not phosphorylate sphingosine. Overexpression increases the formation and secretion of LPA, resulting in transactivation of EGFR and activation of the downstream MAPK signaling pathway, leading to increased cell growth.
Specifications
| AGK | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q53H12 | |
| AGK | |
| Recombinant fragment of human Acylglycerol kinase. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| HL1846 | |
| Unconjugated | |
| AGK | |
| 2610037M15Rik; 6720408I04Rik; acylglycerol kinase; acylglycerol kinase, mitochondrial; Agk; AI465370; CATC5; CTRCT38; FLJ10842; hAGK; hsMuLK; MTDPS10; Mulk; multiple substrate lipid kinase; multi-substrate lipid kinase; RGD1562046 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 55750 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction