missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ AgilePulse™ USB Foot Switch Dual Pedal
Click to view available options
Connection Type:
Output Cable
Unit Size:
Each
Description
- Functions in place of the start button on the front of the generator
- Ideal when conducting in vivo/in ovo gene delivery or nuclear transfer experiments when both hands are needed for sample manipulation
- Compatible with most BTX™ generators
Specifications
Specifications
| Format | Footswitch |
| For Use With (Application) | In-Vivo Gene Delivery, in-Ovo Gene Delivery and Nuclear Transfer/Cloning |
| For Use With (Equipment) | ECM™ 830, ECM™ 2001+, Legacy ECM™ 2001, AgilePulse™, and Gemini X2 Generators and MicroJect 1000 A Microinjection Systems |
| Product Line | BTX Electroporation |
| Product Type | Electroporation accessory |
| Description | AgilePulse Foot Pedal |
| Display | Touchscreen |
| Depth (English) | 3.5 in. |
| Depth (Metric) | 8.9 cm |
| Height (English) | 2 in. |
| Show More |
Intended for Research use only. Not for use in diagnostic or clinical procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction