missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ AgilePulse™ MAX Waveform Generator Only
Click to view available options
Connection Type:
Chamber Cable
Unit Size:
Each
Description
Includes a user-friendly, programmable waveform generator with patented AgilePulse™ technology, BTX™ patent-pending large-volume electroporation chamber, and proprietary BTXPress™ Cytoporation Medium T, optimized for large volume electroporation. IQ/OQ Now available upon request.
- Scale-up
- Transfection protocols readily scale-up from standard laboratory cuvettes to large-volume transfection in the AgilePulse™ MAX system.
- Maximal Efficiency with Cytoporation Medium
- BTXPress™ Cytoporation Medium T used with the AgilePulse™ MAX system has been optimized for maximal efficiency with a number of cell lines, including K562, A20, HEK293 and CHO-KI.
- Compatible with a large range of transfectants including DNA, RNA, siRNA, and olignonucleotides.
- Can be directly diluted in complete growth medium for post-electroporation cell culture.
- Simple User Interface
- All controls are operated with the simple touch screen on the front panel.
- Data is quickly retrieved by USB key and can be analyzed for detailed pulse characteristics including pulse and pulse current.
- AgilePulse™ Advantage
- Transfection efficiency and cell viability are enhanced by specialized, programmable electrical pulse waveforms, particularly important for larger polynucleotide delivery such as DNA plasmids.
- The patented AgilePulse™ technology combines a unique sequence of short high-intensity pulses to porate cell membranes, followed by long low-intensity pulses to further drive transfectants into cells via electrophoresis, while maintaining cell viability.
Specifications
Specifications
| For Use With (Application) | CRISPR Transfections, Transfect Cells Such as Bone Marrow to Produce or Replace a Missing Protein, Deliver siRNA to Suppress Gene Expression, Deliver Genes for Permanent Gene Correction, Load Cells with a Drug for Drug Delivery, Cancer Immunotherapy, Transfect Eukaryotic Cells for Protein Production in Bioreactors, Large-Scale Production of Replication-Deficient Viruses |
| For Use With (Equipment) | BTXpress Cytoporation Medium T or Cytoporation Medium T4, large volume electroporation chamber |
| Frequency | 5 kHz to 1 Hz |
| Product Line | BTX Electroporation |
| Product Type | Electroporator |
| Voltage | 50 to 1000 V |
| Description | AgilePulse MAX Waveform Generator Only |
| Display | Touchscreen |
| Depth (English) | 15.7 in. |
| Depth (Metric) | 40 cm |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction
