Learn More
BTX™ AgilePulse Max Adaptor Cable Set
Product Code. 16644862
Click to view available options
Sterility:
Non-sterile
Unit Size:
Each
Product Code. 16644862
Brand: BTX™ 470097
Adaptor Cable Set connects AgilePulse PLUS to AgilePulse MAX Safety Stands and Large Volume Chamber Stands.
  • Cables and adapters are needed to connect an electrofusion/electroporation system to the electrodes and chambers to be used for a particular application.
Specifications
Description Adaptor Cable Set, Agile Pulse Max
Sterility Non-sterile
Connection Type Chamber Cable
Pulse Form Square Wave