missing translation for 'onlineSavingsMsg'
Learn More

BTX™ AgilePulse™ In Vivo IM (Intra-Muscular) System

Product Code. 15487390
Click to view available options
Connection Type:
Electrode Connector Cable
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15487390

Brand: BTX™ 470500N

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

AgilePulse™ In Vivo IM (Intra-Muscular) System provides maximum efficiency DNA and RNA delivery to skin, muscle, and tumor tissues.

  • Used for animal immunization, vaccine development and gene therapy, provides an intra-muscular, intra-dermal or intra-tumor electroporation solution to produce maximum transfection efficiency
  • Simple user interface
  • Pulse Agile Advantage
  • Uniform, reliable electroporation
  • Simple, Fast Treatment

Specifications

For Use With (Application) Vaccine Development, Intra-Dermal Gene Delivery, Intra-Muscular Gene Delivery, Intra-Tumor Gene Delivery
Frequency 5 kHz to 1 Hz
Product Type Electroporation System
Voltage 50 to 1000 V
Description AgilePulse In Vivo IM (Intra-Muscular) System
Display Touchscreen
Depth (English) 15.7 in.
Depth (Metric) 40 cm
Electrode Shape Needle Array
Height (English) 7.9 in.
Height (Metric) 20 cm
Includes AgilePulse™ IM Generator, Handle for Parallel Needle Array (47-0000), 4-needle Parallel Needle Array (47-0045 - 3 Pk.), 6-needle Parallel Needle Array (47-0070 - 3 Pk.) and IM Software
Operating Temperature (English) 50°F to 104°F
Pulse Length Range 0.200 to 1000 ms
Operating Temperature (Metric) 10°C to 40°C
Pulse Width Range 0.050 to 10 ms
Weight (English) 25 lb.
Weight (Metric) 11.3 kg
Width (English) 12.6 in.
Width (Metric) 32 cm
Sterility Non-sterile
Connection Type Electrode Connector Cable
Interface Touch Screen Display, Footswitch
Pulse Form Square Wave
Show More Show Less

Intended for Research use only. Not for use in diagnostic or clinical procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.