missing translation for 'onlineSavingsMsg'
Learn More

AGI-24512, MedChemExpress

Product Code. 16497287
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497287 5 mg 5mg
16465426 100 mg 100mg
16450877 50 mg 50mg
16434578 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497287 Supplier Medchem Express Supplier No. HY1121305mg
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress AGI-24512 is a potent methionine adenosyltransferase 2α (MAT2A) inhibitor, with an IC50 of 8 nM. AGI-24512 triggers DNA damage response. AGI-24512 can block proliferation of MTAP-deleted cancer cells in vitro. AGI-24512 can be used for researching anticancer.

AGI-24512 is a potent methionine adenosyltransferase 2α (MAT2A) inhibitor, with an IC50 of 8 nM. AGI-24512 triggers DNA damage response. AGI-24512 can block proliferation of MTAP-deleted cancer cells in vitro. AGI-24512 can be used for researching anticancer.

Specifications

Chemical Name or Material AGI-24512
CAS 2201066-53-5
Color Gray
Purity Grade Notes Research
Molecular Formula C24H24N4O2
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (312.13 mM; Need ultrasonic)
SMILES O=C1C(C2=CC=C(O)C=C2)=C(C)NC3=C(N4CCCCC4)C(C5=CC=CC=C5)=NN13
Molecular Weight (g/mol) 400.47
Formula Weight 400.47
Percent Purity 99.48%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.