missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aggrecan Neoepitope Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 19 publications
£186.00 - £403.00
Specifications
| Antigen | Aggrecan Neoepitope |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry reported in scientific literature (PMID 21482542), Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunohistochemistry-Paraffin 1:1000, Immunohistochemistry-Frozen reported in scientific literature (PMID 21482542) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18006164
|
Novus Biologicals
NB100-74350SS |
0.025 mL |
£186.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18775743
|
Novus Biologicals
NB100-74350 |
0.1 mL | |||||||
Description
Aggrecan Neoepitope Polyclonal specifically detects Aggrecan Neoepitope in Human, Mouse, Rat, Porcine, Bovine, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Aggrecan Neoepitope | |
| Western Blot 1:1000, Flow Cytometry reported in scientific literature (PMID 21482542), Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunohistochemistry-Paraffin 1:1000, Immunohistochemistry-Frozen reported in scientific literature (PMID 21482542) | |
| Polyclonal | |
| Rabbit | |
| Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| AGC1SEDK, aggrecan, aggrecan core protein, Cartilage-specific proteoglycan core protein, Chondroitin sulfate proteoglycan 1, Chondroitin sulfate proteoglycan core protein 1, CSPCP, CSPG1aggrecan 1, CSPGCP, large aggregating proteoglycan, MSK16AGCAN | |
| ACAN | |
| IgG | |
| Affinity Purified | |
| Aggrecan Neo (JSCNIT) |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Pig, Bovine, Canine | |
| P16112 | |
| 176 | |
| A synthetic peptide corresponding to a region of human Aggrecan (within residues 350-400). [UniProt# P16112] | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| 61 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title