Learn More
Invitrogen™ Aggrecan Neo Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA11746
Description
PA1-1746 detects human, rat, mouse, bovine, canine, and porcine aggrecan neo G1. PA1-1746 has been successfully used in Western blot and immunofluorescence procedures. By Western blot, this antibody detects an ~64 kDa protein representing the neoepitope sequence (NITEGE) generated by aggrecanase-mediated cleavage at Glu373-Ala374 of aggrecan core proteins. The PA1-1746 immunogen is a synthetic peptide corresponding to residues residues C G G N(387) I T E G E(392) of human aggrecan.
Aggrecan also known as cartilage-specific proteoglycan core protein (CSPCP) or chondroitin sulfate proteoglycan 1 is a protein that is encoded by the ACAN gene. The encoded protein is an integral part of the extracellular matrix in cartilaginous tissue and it withstands compression in cartilage. The human form of the protein is 2316 amino acids long (approximately 250 kDa) and can be expressed in multiple isoforms due to alternative splicing. Aggrecan exhibits a bottlebrush structure, in which chondroitin sulfate and keratan sulfate chains are attached to an extended protein core. The synthesis and degradation of aggrecan are being investigated for their roles in cartilage deterioration during joint injury, disease, and aging. It contains three interglobular domains (IGDs), G1, G2, and G3 that are involved in aggregation, hyaluronan binding, cell adhesion, and chondrocyte apoptosis. Aggrecan fragments from articular cartilage are released into the synovial fluid at all stages of human osteoarthritis as a result of cleavage by aggrecanase and MMP activity. Antibodies BC-3 and BC-4 can be used to detect these neo epitopes of Aggrecanase (ARGSVXXX) and MMP (XXXVDIPEN), respectively.
Specifications
| Aggrecan Neo | |
| Polyclonal | |
| Unconjugated | |
| ACAN | |
| Acan; Agc; AGC1; AGCAN; aggrecan; aggrecan 1; aggrecan 1 (chondroitin sulfate proteoglycan 1, large aggregating proteoglycan, antigen identified by monoclonal antibody A0122); aggrecan core protein; Aggrecan core protein 2; aggrecan CS2 domain; aggrecan epidermal growth factor-like domain-1; aggrecan G2 domain; aggrecan interglobular domain; aggrecan keratan sulfate domain; aggrecan, structural proteoglycan of cartilage; articular cartilage aggrecan precursor; b2b183Clo; cartilage aggregating proteoglycan; cartilage-specific proteoglycan core protein; Chondroitin sulfate proteoglycan 1; chondroitin sulfate proteoglycan core protein 1; cmd; CSPCP; CSPG1; CSPGCP; high density cartilage proteoglycan; large aggregating proteoglycan; large aggregating proteoglycan, antigen; MSK16; SEDK | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11595, 176, 280985, 397255, 403828, 58968 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.83 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide | |
| P07897, P13608, P16112, Q28343, Q29011, Q61282 | |
| ACAN | |
| Synthetic peptide corresponding to residues C G G N(387) I T E G E(392) of human aggrecan. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Bovine, Pig | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.