missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGGF1, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova H00055109-D01P.100ug
This item is not returnable.
View return policy
Description
Sequence: MASEAPSPPRSPPPPTSPEPELAQLRRKVEKLERELRSCKRQVREIEKLLHHTERLYQNAESNNQELRTQVEELSKILQRGRNEDNKKSDVEVQTENHAPWSISDYFYQTYYNDVSLPNKVTELSDQQDQAIETSILNSKDHLQVENDAYPGTDRTENVKYRQVDHFASNSQEPASALATEDTSLEGSSLAESLRAAAEAAVSQTGFSYDENTGLYFDHSTGFYYDSENQLYYDPSTGIYYYCDVESGRYQFHSRVDLQPYPTSSTKQSKDKKLKKKRKDPDSSATNEEKDLNSEDQKAFSVEHTSCNEEENFANMKKKAKIGIHHKNSPPKVTVPTSGNTIESPLHENISNSTSFKDEKIMETDSEPEEGEITDSQTEDSYDEAITSEGNVTAEDSEDEDEDKIWPPCIRVIVIRSPVLQIGSLFIITAVNPATIGREKDMEHTLRIPEVGVSKFHAEIYFDHDLQSYVLVDQGSQNGTIVNGKQILQPKTKCDPYVLEHGDEVKIGETVLSFHIHPGSDTCDGCERGQVRAHLRLDKKDESFVGPTLSKEEKELEGRKELKKIRVKYGLQNTEYEDEKTLKNPKYKDRAGKRREQVGSEGTFQRDDAPASVHSEITDSNKGRKLLEKMGWKKGEGLGKDGGGMKTPIQLQLRRTHAGLGTGKPSSFEDVHLLQNKNKKNWDKARERFTENFPETKPQKDDPGTMPWVKGTLESpecifications
| AGGF1 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| BC032844.1 | |
| AGGF1 | |
| AGGF1 (AAH32844.1, 1 a.a. ∼ 714 a.a) full-length human protein. | |
| RUO | |
| 55109 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| angiogenic factor with G patch and FHA domains 1 | |
| FLJ10283/GPATC7/GPATCH7/HSU84971/HUS84971/VG5Q | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction