missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AGFG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530234
This item is not returnable.
View return policy
Description
Recommended positive controls: IMR32. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (88%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HRB is related to nucleoporins, a class of proteins that mediate nucleocytoplasmic transport. HRB binds the activation domain of the human immunodeficiency virus Rev protein when Rev is assembled onto its RNA target, and is required for the nuclear export of Rev-directed RNAs. The protein encoded by this gene is related to nucleoporins, a class of proteins that mediate nucleocytoplasmic transport. This encoded protein binds the Rev activation domain when Rev is assembled onto its RNA target and can significantly enhance Rev activity when overexpressed. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
Specifications
| AGFG1 | |
| Polyclonal | |
| Unconjugated | |
| AGFG1 | |
| 0710001B06Rik; AGFG1; Agfg1-ps1; AI838505; arf-GAP domain and FG repeat-containing protein 1; arf-GAP domain and FG repeats-containing protein 1; arf-GAP domain and FG repeats-containing protein 1; arf-GAP domain and FG repeat-containing protein 1; ArfGAP with FG repeats 1; AU045498; C130049H11Rik; C85612; D730048C23Rik; HIV-1 Rev binding protein; HIV-1 Rev-binding protein; HIV-1 Rev-binding protein homolog; Hrb; hRIP, Rev interacting protein; MGC116938; MGC116940; NEUG(55-78); Neurogranin; neurogranin (protein kinase C substrate, RC3); Ng; NG/RC3; Nrgn; Nucleoporin-like protein RIP; postsynaptic spine 1; protein kinase C substrate; Pss1; R75334; RAB; Rab, Rev/Rex activation domain-binding protein; RC3; rev/Rex activation domain-binding protein; Rev-interacting protein; Rip; Unknown (protein for MGC:128795) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 15463, 3267 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| P52594, Q8K2K6 | |
| AGFG1 | |
| Recombinant protein encompassing a sequence within the center region of human RIP/Rab. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction