missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGER Mouse anti-Human, Clone: 2A11, Novus Biologicals™
Mouse Monoclonal Antibody
£210.00 - £463.00
Specifications
| Antigen | AGER |
|---|---|
| Clone | 2A11 |
| Dilution | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Surface Plasmon Resonance |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18328205
|
Novus Biologicals
NBP3-11993-0.025MG |
0.025 mg |
£210.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18381556
|
Novus Biologicals
NBP3-11993-0.2MG |
0.2 mg |
£463.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AGER Monoclonal antibody specifically detects AGER in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, ImmunofluorescenceSpecifications
| AGER | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Surface Plasmon Resonance | |
| Unconjugated | |
| RUO | |
| Human | |
| advanced glycosylation end product-specific receptor, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products | |
| Human AGER protein. | |
| Primary |
| 2A11 | |
| Monoclonal | |
| Mouse | |
| Cancer | |
| PBS | |
| 177 | |
| IgG1 κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title