missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGER Antibody (XT-M4) [Janelia Fluor™ 525], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28638JF525
This item is not returnable.
View return policy
Description
AGER Monoclonal antibody specifically detects AGER in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| AGER | |
| Monoclonal | |
| Janelia Fluor 525 | |
| advanced glycosylation end product-specific receptor, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products | |
| AGER/ RAGE | |
| 0.1 mL | |
| Cancer | |
| 177 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| XT-M4 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction