missing translation for 'onlineSavingsMsg'
Learn More

AGER Antibody (2A11), DyLight 650, Novus Biologicals™

Product Code. 18354622
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354622 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18354622 Supplier Bio-Techne Supplier No. NBP311999C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

AGER Monoclonal antibody specifically detects AGER in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGER
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 2A11
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias advanced glycosylation end product-specific receptor, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products
Host Species Rabbit
Immunogen Human AGER protein.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 177
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.