missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AGBL5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550310
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total AGBL5 protein.
AGBL5 has a function in the processing of cytosolic proteins such as alpha tubulin, which is known to be modified by the removal of a C terminal tyrosine. It is expressed in the brain. There are three named isoforms.
Specifications
| AGBL5 | |
| Polyclonal | |
| Unconjugated | |
| AGBL5 | |
| 4930455N08; 9430057O19Rik; Agbl5; ATP/GTP binding protein like 5; ATP/GTP binding protein-like 5; ATP/GTP-binding protein-like 5; carboxypeptidase 5, cytosolic; CCP5; cytosolic carboxypeptidase 5; cytosolic carboxypeptidase-like protein 5; RP75 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 60509 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.1 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q8NDL9 | |
| AGBL5 | |
| Fusion protein corresponding to residues near the C terminal of human ATP/GTP binding protein-like 5. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction