missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGBL3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16990-1-AP-20UL
This item is not returnable.
View return policy
Description
This protein belongs to the peptidase M14 family. It has a function in the processing of cytosolic proteins such as alpha-tubulin, which is known to be modified by the removal of a C-terminal tyrosine. Every subunit binds 1 zinc ion. It has high levels expression in digestive and blood/hematopoietic system, like intestine, urinary, kidney and bone marrow.Specifications
| AGBL3 | |
| Polyclonal | |
| Unconjugated | |
| AGBL3 | |
| AGBL3, ATP/GTP binding protein like 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 340351, 500076, 76223 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8CDP0, Q8NEM8 | |
| AGBL3 | |
| AGBL3 Fusion Protein Ag10592 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction