missing translation for 'onlineSavingsMsg'
Learn More

Agarose, Separation ≥500 bp, Genetic Performance Certified™, Ultrapure

Product Code. 15885218 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 g
250 g
500 g
Packaging:
Plastic Bottle
Unit Size:
100g
250g
500g
This item is not returnable. View return policy

Product Code. 15885218

Brand: Thermo Scientific J75817.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For PCR, DNA/RNA analytical separation, preparative DNA/RNA recovery, DNA/RNA in-gel recovery, and blotting | CAS: 9012-36-6

Recommended for PCR, DNA/RNA analytical separation, preparative DNA/RNA recovery, DNA/RNA in-gel recovery, and blotting

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294

Specifications

Physical Form Granular
Quantity 250 g
Packaging Plastic Bottle
Grade Molecular Biology
Chemical Name or Material Agarose

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.