Learn More
Cytiva Agarose NA Media
Highly purified agarose for separation of nucleic acids to be used as substrates for enzymes. Cytiva™ Agarose NA Media with low electroendosmosis (EEO) is recommended for most DNA/RNA applications.
Brand: Cytiva 17055401
Code : GH
Additional Details : CAS Number : 79-06-1 Weight : 0.02800kg
Description
- Gelling temp 34 to 37°C
Specifications
White | |
Solid | |
Agarose | |
10 g |
Agarose | |
DNA/RNA application | |
60° to 90°C |