missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Agarose, MP Biomedicals™

Product Code. 11442940 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11442940 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11442940 Supplier MP Biomedicals™ Supplier No. 0210026780
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Suitable for both analytical and preparative electrophoresis procedures. Agarose, MP Biomedicals is a purified linear galactan hydrocolloid isolated from agar or agar-bearing algae. It is specially prepared to recover active DNA after separation, with min

This is a standard agarose with good gel strength and clarity, suitable for all general applications.

  • Useful in separation of nucleic acids electrophoretically, because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations
  • Useful in chromatographic separations by either beading the agarose or cross-linking it
  • Agarose gels are used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigen reactions

Specifications

Quantity 100 g
Product Type Agarose Gel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.