missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose ME, for electrophoresis of macromolecules

Product Code. 11472688
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472688 5 g 5g
11482688 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11472688 Supplier Thermo Scientific Chemicals Supplier No. A16693.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9012-36-6

Agarose is frequently used in molecular biology for the separation of large molecules, especially DNA, by electrophoresis. Slabs of agarose gels (usually 0.7 - 2%) for electrophoresis are readily prepared by pouring the warm, liquid solution into a mold. A wide range of different agaroses, of varying molecular weights and properties are commercially available for this purpose. Used in high resolution protein electrophoresis, immunoelectrophoresis, electro immunoassays such as rocket assays, counter current electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Agarose is frequently used in molecular biology for the separation of large molecules, especially DNA, by electrophoresis. Slabs of agarose gels (usually 0.7 - 2%) for electrophoresis are readily prepared by pouring the warm, liquid solution into a mold. A wide range of different agaroses, of varying molecular weights and properties are commercially available for this purpose. Used in high resolution protein electrophoresis, immunoelectrophoresis, electro immunoassays such as rocket assays, counter current electrophoresis.

Solubility
Soluble in water (10 g/L at 80°C).

Notes
Gel point 36°. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, Air, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294

Specifications

Quantity 5 g
Solubility Information Soluble in water (10g/L at 80°C).
Chemical Name or Material Agarose ME, for electrophoresis of macromolecules

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.